Assay Details
Target Gene Details
Entrez Gene ID: | 100529144 |
Gene Name: | CORO7-PAM16 readthrough |
Gene Aliases: |
- |
Location: |
Chr.16:4340251-4416961 on Build GRCh38 |
Assay Gene Location: | Within Intron 27 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CORO7-PAM16 | NM_001201479.1 | NP_001188408.1 | ||
AL833954.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv471075 | Chr.16:4317729 - 4395326 on Build GRCh38 | Loss | CORO7-PAM16 GLIS2-AS1 VASN CORO7 PAM16 GLIS2 |
esv2659570 | Chr.16:4353172 - 4362927 on Build GRCh38 | Deletion | CORO7-PAM16 CORO7 |
esv3637696 | Chr.16:4353267 - 4362846 on Build GRCh38 | Loss | CORO7-PAM16 CORO7 |
nsv1046986 | Chr.16:4049000 - 5555721 on Build GRCh38 | Gain | CORO7-PAM16 RBFOX1 UBALD1 GLIS2 LOC105371063 MIR6769A NUDT16L1 C16orf89 ALG1 SEC14L5 LINC01569 PAM16 CDIP1 C16orf71 NAGPA-AS1 NMRAL1 VASN CORO7 TFAP4 DNAJA3 HMOX2 ROGDI SEPT12 SRL ADCY9 MGRN1 GLYR1 C16orf96 PPL GLIS2-AS1 ANKS3 SMIM22 EEF2KMT NAGPA UBN1 ZNF500 |
nsv571274 | Chr.16:4309420 - 4441189 on Build GRCh38 | Gain | CORO7-PAM16 GLIS2-AS1 VASN CORO7 DNAJA3 PAM16 GLIS2 |
nsv527866 | Chr.16:4206462 - 4412618 on Build GRCh38 | Loss | CORO7-PAM16 GLIS2-AS1 VASN CORO7 TFAP4 LINC01569 PAM16 GLIS2 LOC105371063 SRL |
esv3584924 | Chr.16:4330628 - 4469258 on Build GRCh38 | Gain | CORO7-PAM16 NMRAL1 VASN CORO7 DNAJA3 PAM16 GLIS2 |
esv23732 | Chr.16:4340813 - 4476244 on Build GRCh38 | Gain | CORO7-PAM16 NMRAL1 VASN CORO7 DNAJA3 PAM16 HMOX2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |