Assay Details
Target Gene Details
Entrez Gene ID: | 10595 |
Gene Name: | endoplasmic reticulum to nucleus signaling 2 |
Gene Aliases: |
IRE1-BETA, IRE1b, IRE2p, hIRE2p |
Location: |
Chr.16:23690304-23713500 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ERN2 | NM_001308220.1 | NP_001295149.2 | ||
NM_033266.3 | NP_150296.4 | |||
XM_011545708.1 | XP_011544010.1 | |||
XM_011545709.1 | XP_011544011.1 | |||
XM_011545711.2 | XP_011544013.1 | |||
XM_011545712.2 | XP_011544014.1 | |||
XM_011545713.1 | XP_011544015.1 | |||
AB047079.1 | BAB21297.1 | |||
AK131280.1 | BAD18455.1 | |||
EF560720.1 | ||||
EF560739.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |