Assay Details
Target Gene Details
Entrez Gene ID: | 1001 |
Gene Name: | cadherin 3 |
Gene Aliases: |
CDHP, HJMD, PCAD |
Location: |
Chr.16:68644820-68727573 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CDH3 | NM_001317195.1 | NP_001304124.1 | ||
NM_001317196.1 | NP_001304125.1 | |||
NM_001793.5 | NP_001784.2 | |||
XM_011522800.2 | XP_011521102.1 | |||
AK225396.1 | ||||
AK296968.1 | ||||
AK312554.1 | ||||
BC014462.1 | ||||
BC041846.1 | AAH41846.1 | |||
X63629.1 | CAA45177.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833267 | Chr.16:68648596 - 68823901 on Build GRCh38 | Gain | CDH1 CDH3 |
nsv457515 | Chr.16:68564104 - 68890598 on Build GRCh38 | Gain | CDH1 ZFP90 TANGO6 CDH3 |
nsv572909 | Chr.16:68607485 - 68678827 on Build GRCh38 | Gain | CDH3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |