Assay Details
Target Gene Details
Entrez Gene ID: | 92017 |
Gene Name: | sorting nexin 29 |
Gene Aliases: |
A-388D4.1, RUNDC2A |
Location: |
Chr.16:11976734-12574289 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SNX29 | NM_032167.4 | NP_115543.3 | ||
XM_005255682.3 | XP_005255739.1 | |||
XM_005255683.3 | XP_005255740.1 | |||
XM_011522738.2 | XP_011521040.1 | |||
XM_011522741.2 | XP_011521043.1 | |||
XM_011522743.2 | XP_011521045.1 | |||
XM_011522744.2 | XP_011521046.1 | |||
XM_017023871.1 | XP_016879360.1 | |||
XM_017023872.1 | XP_016879361.1 | |||
XM_017023873.1 | XP_016879362.1 | |||
XM_017023874.1 | XP_016879363.1 | |||
XM_017023876.1 | XP_016879365.1 | |||
XM_017023877.1 | XP_016879366.1 | |||
AK022425.1 | BAB14033.1 | |||
AL135642.1 | ||||
HY078299.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1043959 | Chr.16:11945489 - 12191412 on Build GRCh38 | Gain | LOC642696 TNFRSF17 SNX29 |
nsv571458 | Chr.16:11831151 - 11993425 on Build GRCh38 | Gain | GSPT1 RSL1D1 TNFRSF17 LOC729978 SNX29 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |