Assay Details
Target Gene Details
Entrez Gene ID: | 124446 |
Gene Name: | transmembrane protein 219 |
Gene Aliases: |
IGFBP-3R |
Location: |
Chr.16:29962030-29973052 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMEM219 | NM_001083613.1 | NP_001077082.1 | ||
NM_194280.3 | NP_919256.1 | |||
XM_005255123.1 | XP_005255180.1 | |||
BC017488.1 | AAH17488.1 | |||
BC050051.1 | AAH50051.1 | |||
FJ748884.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3638377 | Chr.16:29967685 - 29971010 on Build GRCh38 | Loss | TMEM219 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |