Assay Details
Target Gene Details
Entrez Gene ID: | 4094 |
Gene Name: | MAF bZIP transcription factor |
Gene Aliases: |
AYGRP, CCA4, CTRCT21, c-MAF |
Location: |
Chr.16:79202624-79600725 on Build GRCh38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MAF |
Target Gene Details
Entrez Gene ID: | 51741 |
Gene Name: | WW domain containing oxidoreductase |
Gene Aliases: |
D16S432E, EIEE28, FOR, FRA16D, HHCMA56, PRO0128, SCAR12, SDR41C1, WOX1 |
Location: |
Chr.16:78099413-79212667 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
WWOX | NM_001291997.1 | NP_001278926.1 | ||
NM_016373.3 | NP_057457.1 | |||
XM_011523103.2 | XP_011521405.1 | |||
XM_017023279.1 | XP_016878768.1 | |||
AF211943.1 | AAF27049.1 | |||
AF227527.1 | AAF82054.1 | |||
AF395123.1 | AAK81727.1 | |||
AF395124.1 | AAK81728.1 | |||
AK027626.1 | ||||
AK290438.1 | ||||
AK291300.1 | ||||
AK298322.1 | ||||
AY256821.1 | AAP94227.1 | |||
BC003184.1 | AAH03184.1 | |||
U13395.1 | AAA21465.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv573287 | Chr.16:79169293 - 79234387 on Build GRCh38 | Loss | MAF LOC105371352 WWOX |
nsv1060491 | Chr.16:79171946 - 79205069 on Build GRCh38 | Loss | MAF LOC105371352 WWOX |
nsv573285 | Chr.16:79124285 - 80093429 on Build GRCh38 | Gain | MAF LOC105371352 LINC01229 MAFTRR WWOX |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |