Assay Details
Target Gene Details
Entrez Gene ID: | 116028 |
Gene Name: | RecQ mediated genome instability 2 |
Gene Aliases: |
BLAP18, C16orf75 |
Location: |
Chr.16:11345438-11351763 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RMI2 | NM_152308.2 | NP_689521.1 | ||
NR_130754.1 | ||||
AK123764.1 | ||||
BC013040.2 | AAH13040.2 | |||
BC022427.1 | AAH22427.1 | |||
BC031016.1 | AAH31016.1 | |||
BC039361.1 | AAH39361.1 | |||
CN311350.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv2728n100 | Chr.16:11333115 - 11449038 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv1040428 | Chr.16:10145408 - 11362136 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)