Assay Details
Target Gene Details
Entrez Gene ID: | 729993 |
Gene Name: | shisa family member 9 |
Gene Aliases: |
CKAMP44 |
Location: |
Chr.16:12901619-13563427 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SHISA9 | NM_001145204.2 | NP_001138676.2 | ||
XM_005255539.3 | XP_005255596.2 | |||
XM_011522642.2 | XP_011520944.1 | |||
AK299610.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833142 | Chr.16:12828715 - 13005564 on Build GRCh38 | Loss | SHISA9 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |