Assay Details
Target Gene Details
Entrez Gene ID: | 5336 |
Gene Name: | phospholipase C gamma 2 |
Gene Aliases: |
APLAID, FCAS3, PLC-IV, PLC-gamma-2 |
Location: |
Chr.16:81779258-81962693 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PLCG2 | NM_002661.4 | NP_002652.2 | ||
AB208914.1 | BAD92151.1 | |||
BC007565.1 | AAH07565.1 | |||
BC011772.2 | AAH11772.1 | |||
BC014561.2 | AAH14561.1 | |||
BC018646.2 | AAH18646.1 | |||
BP351485.1 | ||||
DB456860.1 | ||||
M37238.1 | AAA60112.1 | |||
X14034.1 | CAA32194.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv573365 | Chr.16:81771367 - 81942287 on Build GRCh38 | Loss | LOC105369213 PLCG2 |
esv3639343 | Chr.16:81755626 - 81815373 on Build GRCh38 | Loss | LOC105369213 PLCG2 |
esv3639339 | Chr.16:81696019 - 81869162 on Build GRCh38 | Loss | LOC105369213 CMIP PLCG2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |