Assay Details
Target Gene Details
Entrez Gene ID: | 100529144 |
Gene Name: | CORO7-PAM16 readthrough |
Gene Aliases: |
- |
Location: |
Chr.16:4340251-4416961 on Build GRCh38 |
Assay Gene Location: | Within Intron 27 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CORO7-PAM16 | NM_001201479.1 | NP_001188408.1 | ||
AL833954.1 |
Target Gene Details
Entrez Gene ID: | 51025 |
Gene Name: | presequence translocase-associated motor 16 homolog (S. cerevisiae) |
Gene Aliases: |
CGI-136, MAGMAS, SMDMDM, TIM16, TIMM16 |
Location: |
Chr.16:4340251-4351372 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PAM16 | NM_016069.9 | NP_057153.8 | ||
AF151894.1 | AAD34131.1 | |||
AF349455.1 | AAL57767.1 | |||
AK026514.1 | BAB15494.1 | |||
BC005024.1 | AAH05024.1 | |||
BG715429.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv471075 | Chr.16:4317729 - 4395326 on Build GRCh38 | Loss | CORO7 CORO7-PAM16 PAM16 GLIS2-AS1 VASN GLIS2 |
nsv1046986 | Chr.16:4049000 - 5555721 on Build GRCh38 | Gain | PPL C16orf71 ROGDI MIR6769A MGRN1 LINC01569 SRL PAM16 GLIS2-AS1 NMRAL1 VASN ZNF500 CORO7-PAM16 EEF2KMT RBFOX1 HMOX2 ALG1 TFAP4 NAGPA-AS1 ANKS3 LOC105371063 CDIP1 GLIS2 NAGPA UBN1 NUDT16L1 CORO7 C16orf89 SEC14L5 SMIM22 DNAJA3 SEPT12 C16orf96 UBALD1 ADCY9 GLYR1 |
nsv571274 | Chr.16:4309420 - 4441189 on Build GRCh38 | Gain | CORO7 CORO7-PAM16 PAM16 DNAJA3 GLIS2-AS1 VASN GLIS2 |
nsv527866 | Chr.16:4206462 - 4412618 on Build GRCh38 | Loss | CORO7 CORO7-PAM16 LOC105371063 LINC01569 SRL PAM16 GLIS2-AS1 TFAP4 VASN GLIS2 |
esv3584924 | Chr.16:4330628 - 4469258 on Build GRCh38 | Gain | CORO7 CORO7-PAM16 PAM16 DNAJA3 NMRAL1 VASN GLIS2 |
esv23732 | Chr.16:4340813 - 4476244 on Build GRCh38 | Gain | CORO7 CORO7-PAM16 HMOX2 PAM16 DNAJA3 NMRAL1 VASN |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |