Assay Details
Target Gene Details
Entrez Gene ID: | 1428 |
Gene Name: | crystallin mu |
Gene Aliases: |
DFNA40, THBP |
Location: |
Chr.16:21258518-21303136 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CRYM | NM_001888.4 | NP_001879.1 | ||
AK290852.1 | ||||
AK310424.1 | ||||
BC018061.1 | AAH18061.1 | |||
BI860684.1 | ||||
BX648477.1 | CAI46030.1 | |||
L02950.1 | AAC16914.1 | |||
U85772.1 | AAB81564.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3638165 | Chr.16:21188383 - 21267495 on Build GRCh38 | Gain | ANKS4B CRYM ZP2 |
nsv571623 | Chr.16:21084049 - 21341186 on Build GRCh38 | Gain | DNAH3 TMEM159 CRYM-AS1 ANKS4B CRYM ZP2 |
esv3638164 | Chr.16:21185596 - 21338776 on Build GRCh38 | Gain | CRYM-AS1 ANKS4B CRYM ZP2 |
nsv436835 | Chr.16:18930652 - 21500976 on Build GRCh38 | Insertion | SMG1P3 GDE1 COQ7 DNAH3 ACSM5 GPR139 TMEM159 C16orf62 SNX29P1 TMC5 LOC102723385 TMC7 IQCK ACSM2B CCP110 LOC81691 KNOP1 LYRM1 THUMPD1 UMOD DCUN1D3 GPRC5B CRYM-AS1 SYT17 GP2 ANKS4B PDILT ACSM2A ACSM1 LOC100190986 ITPRIPL2 ERI2 NPIPB3 CLEC19A CRYM ACSM3 ZP2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |