Assay Details
Target Gene Details
Entrez Gene ID: | 9074 |
Gene Name: | claudin 6 |
Gene Aliases: |
- |
Location: |
Chr.16:3014712-3018187 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CLDN6 | NM_021195.4 | NP_067018.2 | ||
AK075329.1 | ||||
BX444798.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv509588 | Chr.16:2919880 - 3043776 on Build GRCh38 | Insertion | THOC6 LINC00514 KREMEN2 BICDL2 FLYWCH1 PAQR4 LOC101929613 CLDN9 PKMYT1 TNFRSF12A LOC100128770 HCFC1R1 CLDN6 |
nsv833125 | Chr.16:3017124 - 3131817 on Build GRCh38 | Loss | THOC6 IL32 ZNF205-AS1 ZNF205 BICDL2 ZSCAN10 MMP25 TNFRSF12A ZNF213-AS1 LOC100128770 MMP25-AS1 HCFC1R1 CLDN6 |
nsv952906 | Chr.16:2868600 - 3028099 on Build GRCh38 | Deletion | THOC6 FLYWCH2 LINC00514 KREMEN2 BICDL2 FLYWCH1 PAQR4 LOC101929613 CLDN9 PKMYT1 TNFRSF12A HCFC1R1 CLDN6 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |