Assay Details
Target Gene Details
Entrez Gene ID: | 23295 |
Gene Name: | mahogunin ring finger 1 |
Gene Aliases: |
RNF156 |
Location: |
Chr.16:4616493-4690974 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MGRN1 | XM_011522441.1 | XP_011520743.1 | ||
XM_017023093.1 | XP_016878582.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1160333 | Chr.16:4570296 - 4709737 on Build GRCh38 | Deletion | C16orf96 MGRN1 NUDT16L1 ANKS3 UBALD1 MIR6769A |
nsv1046986 | Chr.16:4049000 - 5555721 on Build GRCh38 | Gain | LINC01569 NUDT16L1 CDIP1 ZNF500 PPL UBALD1 TFAP4 SRL EEF2KMT C16orf71 NAGPA-AS1 SMIM22 NMRAL1 ALG1 ANKS3 RBFOX1 CORO7-PAM16 VASN GLIS2-AS1 CORO7 LOC105371063 SEC14L5 PAM16 GLIS2 UBN1 HMOX2 ADCY9 MIR6769A GLYR1 C16orf96 DNAJA3 C16orf89 MGRN1 NAGPA SEPT12 ROGDI |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |