Assay Details
Target Gene Details
Entrez Gene ID: | 60490 |
Gene Name: | phosphopantothenoylcysteine decarboxylase |
Gene Aliases: |
MDS018, PPC-DC, coaC |
Location: |
Chr.15:75023533-75060180 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PPCDC | NM_001301101.1 | NP_001288030.1 | ||
NM_001301102.1 | NP_001288031.1 | |||
NM_001301103.1 | NP_001288032.1 | |||
NM_021823.4 | NP_068595.3 | |||
NR_125369.1 | ||||
XM_017022461.1 | XP_016877950.1 | |||
XM_017022464.1 | XP_016877953.1 | |||
XM_017022465.1 | XP_016877954.1 | |||
AK027491.1 | BAB55151.1 | |||
AY358848.1 | AAQ89207.1 | |||
BC014409.1 | AAH14409.1 | |||
BF243417.1 | ||||
BI833814.1 | ||||
BU500301.1 | ||||
DB488540.1 | ||||
DW437029.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv569993 | Chr.15:74949814 - 75191411 on Build GRCh38 | Gain | SCAMP5 PPCDC RPP25 |
nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion | CYP11A1 LOC107984728 MIR4513 ARID3B SCAMP2 RPP25 HIGD2B CCDC33 C15orf59 STRA6 SCAMP5 UBL7-AS1 LOXL1 ADPGK-AS1 PPCDC FAM219B C15orf59-AS1 REC114 TBC1D21 UBL7 EDC3 ULK3 ISLR2 LOC105376731 CYP1A1 CD276 MIR6882 STOML1 LOC732265 PML LOC729739 COX5A CPLX3 ADPGK LOC101929333 ISLR LMAN1L GOLGA6C MPI SEMA7A HCN4 MIR6881 NEO1 BBS4 NPTN CLK3 C15orf39 CYP1A2 GOLGA6A LOC283731 NPTN-IT1 CSK LOXL1-AS1 LOC102723750 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |