Assay Details
Target Gene Details
Entrez Gene ID: | 79094 |
Gene Name: | ChaC glutathione specific gamma-glutamylcyclotransferase 1 |
Gene Aliases: |
- |
Location: |
Chr.15:40942135-40956519 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CHAC1 | XM_017022563.1 | XP_016878052.1 | ||
XM_017022564.1 | XP_016878053.1 | |||
XM_017022565.1 | XP_016878054.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1504 | Chr.15:40936351 - 40969374 on Build GRCh38 | Insertion | CHAC1 DLL4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |