Assay Details
Target Gene Details
Entrez Gene ID: | 4756 |
Gene Name: | neogenin 1 |
Gene Aliases: |
IGDCC2, NGN, NTN1R2 |
Location: |
Chr.15:73051715-73305206 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion | RPP25 STRA6 NEO1 ADPGK-AS1 NPTN COX5A GOLGA6C LOXL1 NPTN-IT1 CYP1A1 LOC732265 CPLX3 STOML1 LOC283731 SCAMP5 HIGD2B REC114 ISLR2 CYP11A1 LOXL1-AS1 EDC3 PPCDC UBL7-AS1 MIR4513 ADPGK MIR6882 FAM219B MPI GOLGA6A PML CD276 BBS4 LOC105376731 MIR6881 TBC1D21 C15orf59 ULK3 ARID3B CSK LOC729739 SCAMP2 ISLR C15orf39 CCDC33 LMAN1L CLK3 LOC101929333 C15orf59-AS1 UBL7 SEMA7A CYP1A2 LOC102723750 HCN4 LOC107984728 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |