Assay Details
Target Gene Details
Entrez Gene ID: | 4756 |
Gene Name: | neogenin 1 |
Gene Aliases: |
IGDCC2, NGN, NTN1R2 |
Location: |
Chr.15:73051715-73305206 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv525462 | Chr.15:73057292 - 73271154 on Build GRCh38 | Loss | NEO1 |
nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion | LOC283731 CYP1A2 ADPGK-AS1 LOC101929333 STOML1 LOC102723750 SCAMP5 C15orf59-AS1 LOC729739 ISLR2 MIR4513 LMAN1L CD276 GOLGA6C SCAMP2 MIR6881 FAM219B STRA6 MPI LOC107984728 SEMA7A CYP11A1 REC114 LOXL1-AS1 ADPGK NPTN-IT1 TBC1D21 C15orf39 COX5A ISLR LOC732265 ARID3B C15orf59 CCDC33 UBL7 CPLX3 GOLGA6A HCN4 CYP1A1 EDC3 BBS4 HIGD2B UBL7-AS1 NPTN RPP25 PPCDC CSK LOXL1 ULK3 MIR6882 PML CLK3 LOC105376731 NEO1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |