Assay Details
Target Gene Details
Entrez Gene ID: | 8554 |
Gene Name: | protein inhibitor of activated STAT 1 |
Gene Aliases: |
DDXBP1, GBP, GU/RH-II, ZMIZ3 |
Location: |
Chr.15:68054179-68191464 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PIAS1 | NM_001320687.1 | NP_001307616.1 | ||
NM_016166.2 | NP_057250.1 | |||
XM_011522126.2 | XP_011520428.1 | |||
XM_011522127.2 | XP_011520429.1 | |||
XM_017022688.1 | XP_016878177.1 | |||
XM_017022689.1 | XP_016878178.1 | |||
XM_017022690.1 | XP_016878179.1 | |||
XM_017022691.1 | XP_016878180.1 | |||
AF077951.1 | AAC36702.1 | |||
AF167160.1 | AAD49722.1 | |||
AK094641.1 | ||||
AK314515.1 | ||||
AY826819.1 | ||||
BC118587.1 | ||||
BC121797.1 | ||||
EF487997.1 | ||||
EF487998.1 | ||||
FJ997900.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1037980 | Chr.15:68061238 - 68145365 on Build GRCh38 | Loss | PIAS1 |
nsv517512 | Chr.15:68026786 - 68132927 on Build GRCh38 | Gain | PIAS1 |
nsv569803 | Chr.15:67585107 - 68124237 on Build GRCh38 | Gain | LOC101929076 MAP2K5 SKOR1 PIAS1 RNU6-1 |
nsv521762 | Chr.15:68057555 - 68179553 on Build GRCh38 | Gain | PIAS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |