Assay Details
Target Gene Details
Entrez Gene ID: | 4756 |
Gene Name: | neogenin 1 |
Gene Aliases: |
IGDCC2, NGN, NTN1R2 |
Location: |
Chr.15:73051715-73305206 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv525462 | Chr.15:73057292 - 73271154 on Build GRCh38 | Loss | NEO1 |
nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion | CYP1A2 LOC729739 LOXL1 CCDC33 PPCDC CLK3 LOC107984728 EDC3 SEMA7A ULK3 ADPGK CD276 FAM219B NEO1 CYP1A1 HIGD2B REC114 COX5A LOC105376731 ARID3B SCAMP2 STRA6 LOC732265 ISLR2 NPTN GOLGA6C SCAMP5 CYP11A1 BBS4 LOXL1-AS1 LMAN1L TBC1D21 C15orf59-AS1 MIR4513 RPP25 MIR6881 HCN4 MIR6882 LOC102723750 CPLX3 LOC283731 LOC101929333 C15orf59 CSK PML UBL7 NPTN-IT1 MPI C15orf39 GOLGA6A ISLR ADPGK-AS1 STOML1 UBL7-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |