Assay Details
Target Gene Details
Entrez Gene ID: | 5607 |
Gene Name: | mitogen-activated protein kinase kinase 5 |
Gene Aliases: |
HsT17454, MAPKK5, MEK5, PRKMK5 |
Location: |
Chr.15:67542606-67807117 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MAP2K5 | NM_002757.3 | NP_002748.1 | ||
NM_145160.2 | NP_660143.1 | |||
XM_011521787.2 | XP_011520089.1 | |||
XM_011521788.2 | XP_011520090.1 | |||
XM_017022414.1 | XP_016877903.1 | |||
AK315435.1 | ||||
BC008838.2 | AAH08838.1 | |||
BT006780.1 | AAP35426.1 | |||
CR542229.1 | CAG47025.1 | |||
DN998338.1 | ||||
U71087.1 | AAB16851.1 | |||
U71088.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv569799 | Chr.15:67511204 - 67585107 on Build GRCh38 | Loss | C15orf61 IQCH-AS1 MAP2K5 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |