Assay Details
Target Gene Details
Entrez Gene ID: | 1445 |
Gene Name: | c-src tyrosine kinase |
Gene Aliases: |
- |
Location: |
Chr.15:74782064-74803198 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CSK | NM_001127190.1 | NP_001120662.1 | ||
NM_004383.2 | NP_004374.1 | |||
XM_005254165.4 | XP_005254222.1 | |||
XM_017021925.1 | XP_016877414.1 | |||
AK223626.1 | BAD97346.1 | |||
AK290531.1 | ||||
AK298245.1 | ||||
AK310136.1 | ||||
AK312669.1 | ||||
BC104847.1 | AAI04848.1 | |||
BC104875.1 | AAI04876.1 | |||
BC106073.1 | AAI06074.1 | |||
X59932.1 | CAA42556.1 | |||
X60114.1 | CAA42713.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833055 | Chr.15:74696003 - 74873947 on Build GRCh38 | Loss | CPLX3 CSK EDC3 LMAN1L ULK3 SCAMP2 CYP1A1 MIR6882 MIR4513 CYP1A2 |
nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion | CSK RPP25 MPI NEO1 LOC102723750 CYP11A1 ADPGK-AS1 SCAMP5 CD276 STOML1 COX5A ARID3B NPTN-IT1 HIGD2B CLK3 LOC729739 LOC283731 MIR4513 CPLX3 ISLR2 LMAN1L LOXL1-AS1 MIR6882 STRA6 PML LOXL1 C15orf59-AS1 CCDC33 LOC105376731 TBC1D21 LOC101929333 ULK3 FAM219B CYP1A1 C15orf59 UBL7-AS1 C15orf39 NPTN GOLGA6C UBL7 GOLGA6A REC114 ADPGK EDC3 BBS4 SCAMP2 CYP1A2 PPCDC LOC732265 HCN4 LOC107984728 MIR6881 SEMA7A ISLR |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |