Assay Details
Target Gene Details
Entrez Gene ID: | 3067 |
Gene Name: | histidine decarboxylase |
Gene Aliases: |
- |
Location: |
Chr.15:50241945-50266049 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HDC | NM_001306146.1 | NP_001293075.1 | ||
NM_002112.3 | NP_002103.2 | |||
XM_017022094.1 | XP_016877583.1 | |||
XM_017022095.1 | XP_016877584.1 | |||
XM_017022097.1 | XP_016877586.1 | |||
XM_017022099.1 | XP_016877588.1 | |||
AK308833.1 | ||||
AK310479.1 | ||||
AK311072.1 | ||||
BC130527.1 | ||||
BC144173.1 | ||||
DA916790.1 | ||||
M60445.1 | AAC41698.1 | |||
X54297.1 | CAA38196.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |