Assay Details
Target Gene Details
Entrez Gene ID: | 2558 |
Gene Name: | gamma-aminobutyric acid type A receptor alpha5 subunit |
Gene Aliases: |
- |
Location: |
Chr.15:26866719-26949210 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GABRA5 | NM_000810.3 | NP_000801.1 | ||
NM_001165037.1 | NP_001158509.1 | |||
XM_005268258.2 | XP_005268315.1 | |||
XM_006720459.2 | XP_006720522.1 | |||
XM_017022055.1 | XP_016877544.1 | |||
XM_017022056.1 | XP_016877545.1 | |||
A28104.1 | CAA01920.1 | |||
AK290453.1 | ||||
AK294303.1 | ||||
AK303742.1 | ||||
BC111979.1 | AAI11980.1 | |||
BC113422.1 | ||||
BT009830.1 | AAP88832.1 | |||
L08485.1 | AAA58490.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2668795 | Chr.15:26828644 - 27745218 on Build GRCh38 | Deletion | GABRG3 GABRA5 GABRG3-AS1 OCA2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |