Assay Details
Target Gene Details
Entrez Gene ID: | 8482 |
Gene Name: | semaphorin 7A (John Milton Hagen blood group) |
Gene Aliases: |
CD108, CDw108, H-SEMA-K1, H-Sema-L, JMH, SEMAK1, SEMAL |
Location: |
Chr.15:74409284-74433959 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SEMA7A | NM_001146029.2 | NP_001139501.1 | ||
NM_001146030.2 | NP_001139502.1 | |||
NM_003612.4 | NP_003603.1 | |||
AF030698.1 | AAC34261.1 | |||
AF069493.1 | AAC82642.1 | |||
AF071542.1 | AAC80456.1 | |||
AK293280.1 | ||||
AK312257.1 | ||||
AM180445.1 | CAJ55398.1 | |||
AM180446.1 | CAJ55399.1 | |||
AM180447.1 | CAJ55400.1 | |||
AM180448.1 | CAJ55401.1 | |||
AM180449.1 | CAJ55402.1 | |||
AM180450.1 | CAJ55403.1 | |||
AM180451.1 | CAJ55404.1 | |||
BC101643.1 | AAI01644.1 | |||
BC101647.1 | AAI01648.1 | |||
DA136330.1 | ||||
DA232032.1 | ||||
DA815098.1 | ||||
DC408557.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv569979 | Chr.15:74417064 - 74430583 on Build GRCh38 | Loss |
![]() |
nsv569982 | Chr.15:74417784 - 74429628 on Build GRCh38 | Gain |
![]() |
nsv569974 | Chr.15:74408859 - 74434772 on Build GRCh38 | Gain |
![]() ![]() |
nsv569976 | Chr.15:74416730 - 74422870 on Build GRCh38 | Loss |
![]() |
nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)