Assay Details
Target Gene Details
Entrez Gene ID: | 4756 |
Gene Name: | neogenin 1 |
Gene Aliases: |
IGDCC2, NGN, NTN1R2 |
Location: |
Chr.15:73051715-73305206 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv525462 | Chr.15:73057292 - 73271154 on Build GRCh38 | Loss | NEO1 |
nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion | LOC107984728 C15orf59-AS1 MIR4513 COX5A MPI C15orf39 REC114 SEMA7A LOC729739 CPLX3 LOXL1-AS1 RPP25 NPTN PML C15orf59 CYP11A1 LOC283731 LOC101929333 PPCDC ULK3 CYP1A1 LOC102723750 CD276 ISLR2 CSK UBL7-AS1 CLK3 HIGD2B CYP1A2 ADPGK ADPGK-AS1 NPTN-IT1 SCAMP5 HCN4 EDC3 GOLGA6C MIR6882 ARID3B SCAMP2 CCDC33 FAM219B STRA6 UBL7 NEO1 BBS4 LOXL1 TBC1D21 LOC105376731 LOC732265 ISLR MIR6881 STOML1 GOLGA6A LMAN1L |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |