Assay Details
Target Gene Details
Entrez Gene ID: | 4756 |
Gene Name: | neogenin 1 |
Gene Aliases: |
IGDCC2, NGN, NTN1R2 |
Location: |
Chr.15:73051715-73305206 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv525462 | Chr.15:73057292 - 73271154 on Build GRCh38 | Loss | NEO1 |
nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion | TBC1D21 LOC283731 MPI C15orf59 BBS4 CD276 MIR4513 LOC732265 LOC107984728 LOXL1-AS1 STOML1 ADPGK-AS1 SEMA7A HIGD2B FAM219B NPTN-IT1 CYP11A1 EDC3 SCAMP2 C15orf59-AS1 RPP25 LOC102723750 LOC101929333 HCN4 SCAMP5 CCDC33 CYP1A2 STRA6 ISLR2 UBL7 PPCDC C15orf39 COX5A UBL7-AS1 GOLGA6A GOLGA6C ISLR NEO1 ADPGK CPLX3 PML NPTN REC114 LMAN1L ARID3B CYP1A1 LOXL1 LOC105376731 ULK3 CLK3 LOC729739 CSK MIR6881 MIR6882 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |