Assay Details
Target Gene Details
Entrez Gene ID: | 10260 |
Gene Name: | DENN domain containing 4A |
Gene Aliases: |
IRLB, MYCPBP |
Location: |
Chr.15:65659123-65792293 on Build GRCh38 |
Assay Gene Location: | Within Intron 27 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DENND4A | NM_001144823.2 | NP_001138295.1 | ||
NM_001320835.1 | NP_001307764.1 | |||
NM_005848.4 | NP_005839.3 | |||
XM_005254120.4 | XP_005254177.1 | |||
XM_005254121.3 | XP_005254178.1 | |||
XM_005254122.4 | XP_005254179.1 | |||
XM_006720371.3 | XP_006720434.1 | |||
XM_011521157.2 | XP_011519459.1 | |||
XM_017021862.1 | XP_016877351.1 | |||
XM_017021863.1 | XP_016877352.1 | |||
XM_017021864.1 | XP_016877353.1 | |||
XM_017021865.1 | XP_016877354.1 | |||
AF534403.1 | AAQ10514.1 | |||
AK091268.1 | ||||
AL832602.2 | CAD89960.1 | |||
AL833317.1 | ||||
AL833384.1 | ||||
BC041706.1 | AAH41706.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833043 | Chr.15:65638528 - 65801425 on Build GRCh38 | Loss | MIR4511 LOC646358 SLC24A1 DENND4A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |