Assay Details
Target Gene Details
Entrez Gene ID: | 105371031 |
Gene Name: | translation initiation factor IF-2-like |
Gene Aliases: |
- |
Location: |
Chr.15:89399716-89507395 on Build GRCh38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC105371031 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1038140 | Chr.15:89141274 - 89620108 on Build GRCh38 | Gain | LOC105370962 MIR9-3 RLBP1 LINC00928 ABHD2 MIR6766 POLG MIR9-3HG RHCG FANCI LOC105371031 TICRR |
esv34025 | Chr.15:89412131 - 89630626 on Build GRCh38 | Loss | LINC00928 KIF7 RHCG LOC105371031 TICRR |
nsv833086 | Chr.15:89275748 - 89437378 on Build GRCh38 | Loss | LOC105370962 MIR9-3 MIR6766 POLG MIR9-3HG FANCI LOC105371031 |