Assay Details
Target Gene Details
Entrez Gene ID: | 1445 |
Gene Name: | c-src tyrosine kinase |
Gene Aliases: |
- |
Location: |
Chr.15:74782064-74803198 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CSK | NM_001127190.1 | NP_001120662.1 | ||
NM_004383.2 | NP_004374.1 | |||
XM_005254165.4 | XP_005254222.1 | |||
XM_017021925.1 | XP_016877414.1 | |||
AK223626.1 | BAD97346.1 | |||
AK290531.1 | ||||
AK298245.1 | ||||
AK310136.1 | ||||
AK312669.1 | ||||
BC104847.1 | AAI04848.1 | |||
BC104875.1 | AAI04876.1 | |||
BC106073.1 | AAI06074.1 | |||
X59932.1 | CAA42556.1 | |||
X60114.1 | CAA42713.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833055 | Chr.15:74696003 - 74873947 on Build GRCh38 | Loss | SCAMP2 CSK CYP1A2 MIR4513 CYP1A1 EDC3 LMAN1L ULK3 MIR6882 CPLX3 |
nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion | LOC283731 TBC1D21 CD276 MIR6881 MPI ULK3 BBS4 ADPGK PML CSK COX5A CYP1A2 NPTN NPTN-IT1 ADPGK-AS1 NEO1 LOXL1-AS1 EDC3 LOC732265 ISLR2 LOC105376731 CYP11A1 STRA6 UBL7-AS1 GOLGA6C SCAMP2 RPP25 CLK3 CCDC33 SCAMP5 LOC729739 CYP1A1 LMAN1L C15orf59-AS1 ISLR C15orf39 CPLX3 PPCDC SEMA7A FAM219B REC114 C15orf59 HCN4 STOML1 LOC107984728 MIR4513 ARID3B LOC102723750 LOXL1 LOC101929333 GOLGA6A UBL7 MIR6882 HIGD2B |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |