Assay Details
Target Gene Details
Entrez Gene ID: | 4756 |
Gene Name: | neogenin 1 |
Gene Aliases: |
IGDCC2, NGN, NTN1R2 |
Location: |
Chr.15:73051715-73305206 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv525462 | Chr.15:73057292 - 73271154 on Build GRCh38 | Loss | NEO1 |
nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion | HCN4 PML ULK3 MIR6882 PPCDC SEMA7A LOC732265 FAM219B CCDC33 CYP1A2 C15orf59 NEO1 HIGD2B LMAN1L REC114 LOXL1 LOC105376731 LOC101929333 STRA6 MIR4513 RPP25 LOC102723750 LOXL1-AS1 CYP1A1 LOC107984728 UBL7 ARID3B CSK LOC283731 GOLGA6C NPTN ADPGK ISLR2 COX5A SCAMP5 TBC1D21 ISLR BBS4 MIR6881 EDC3 ADPGK-AS1 STOML1 CPLX3 C15orf39 SCAMP2 C15orf59-AS1 UBL7-AS1 CD276 CLK3 MPI GOLGA6A CYP11A1 LOC729739 NPTN-IT1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |