Assay Details
Target Gene Details
Entrez Gene ID: | 51495 |
Gene Name: | 3-hydroxyacyl-CoA dehydratase 3 |
Gene Aliases: |
B-IND1, BIND1, HSPC121, PTPLAD1 |
Location: |
Chr.15:65530489-65578355 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HACD3 | NM_016395.2 | NP_057479.2 | ||
XM_011521664.1 | XP_011519966.1 | |||
AF161470.1 | AAF29085.1 | |||
AJ271091.1 | CAB69070.1 | |||
AK027421.1 | BAB55101.1 | |||
AK074898.1 | BAC11277.1 | |||
AK225120.1 | ||||
AK299234.1 | ||||
BC035508.1 | AAH35508.1 | |||
BC047685.1 | AAH47685.1 | |||
BC058912.1 | AAH58912.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833042 | Chr.15:65446079 - 65656395 on Build GRCh38 | Loss | VWA9 HACD3 SLC24A1 DPP8 |
nsv569787 | Chr.15:65525552 - 65538180 on Build GRCh38 | Loss | HACD3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |