Assay Details
Target Gene Details
Entrez Gene ID: | 4756 |
Gene Name: | neogenin 1 |
Gene Aliases: |
IGDCC2, NGN, NTN1R2 |
Location: |
Chr.15:73051715-73305206 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv525462 | Chr.15:73057292 - 73271154 on Build GRCh38 | Loss | NEO1 |
nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion | MPI LOC101929333 C15orf39 CSK GOLGA6A CYP1A1 CYP11A1 NPTN LOXL1-AS1 LOXL1 ARID3B CD276 STRA6 ISLR CYP1A2 LOC105376731 HIGD2B TBC1D21 GOLGA6C EDC3 RPP25 CCDC33 REC114 C15orf59-AS1 C15orf59 HCN4 LOC729739 UBL7 PPCDC SCAMP2 FAM219B PML SCAMP5 BBS4 LOC107984728 MIR6882 NPTN-IT1 LOC283731 SEMA7A ISLR2 ULK3 ADPGK NEO1 CPLX3 MIR6881 CLK3 STOML1 LOC102723750 ADPGK-AS1 UBL7-AS1 LMAN1L MIR4513 LOC732265 COX5A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |