Assay Details
Target Gene Details
Entrez Gene ID: | 89978 |
Gene Name: | diphthamine biosynthesis 6 |
Gene Aliases: |
ATPBD4 |
Location: |
Chr.15:35217356-35546222 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DPH6 | XM_017022708.1 | XP_016878197.1 | ||
BX537862.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3636131 | Chr.15:35225355 - 35227584 on Build GRCh38 | Loss | DPH6 |
nsv1486 | Chr.15:35202047 - 35244348 on Build GRCh38 | Deletion | DPH6 ANP32AP1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |