Assay Details
Target Gene Details
Entrez Gene ID: | 51621 |
Gene Name: | Kruppel like factor 13 |
Gene Aliases: |
BTEB3, FKLF2, NSLP1, RFLAT-1, RFLAT1 |
Location: |
Chr.15:31326855-31435665 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KLF13 | NM_001302461.1 | NP_001289390.1 | ||
NM_015995.3 | NP_057079.2 | |||
AF132599.1 | AAD26864.1 | |||
AF150628.1 | AAD34020.1 | |||
AI084187.1 | ||||
BC010438.1 | AAH10438.1 | |||
BC012741.2 | AAH12741.1 | |||
BC013946.2 | AAH13946.1 | |||
KJ534876.1 | ||||
KJ535015.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3636052 | Chr.15:31325578 - 31378949 on Build GRCh38 | Gain | KLF13 |
nsv516681 | Chr.15:31324471 - 31383250 on Build GRCh38 | Loss | KLF13 |
nsv456748 | Chr.15:31329118 - 31405439 on Build GRCh38 | Loss | LOC105370939 KLF13 |
esv34100 | Chr.15:31204311 - 31500388 on Build GRCh38 | Loss | LOC105370939 OTUD7A LOC283710 KLF13 LOC102725022 |
nsv568772 | Chr.15:31245293 - 31345793 on Build GRCh38 | Loss | KLF13 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |