Assay Details
Target Gene Details
Entrez Gene ID: | 4756 |
Gene Name: | neogenin 1 |
Gene Aliases: |
IGDCC2, NGN, NTN1R2 |
Location: |
Chr.15:73051715-73305206 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv525462 | Chr.15:73057292 - 73271154 on Build GRCh38 | Loss | NEO1 |
nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion | LOC101929333 SEMA7A REC114 SCAMP5 CPLX3 GOLGA6C LMAN1L CLK3 CSK RPP25 ARID3B LOXL1-AS1 LOC729739 CYP1A1 C15orf59-AS1 LOC732265 MPI EDC3 NPTN-IT1 HCN4 STRA6 ISLR TBC1D21 STOML1 CCDC33 MIR4513 UBL7-AS1 ISLR2 LOC105376731 C15orf39 SCAMP2 CD276 LOXL1 ADPGK-AS1 LOC102723750 ULK3 MIR6882 FAM219B HIGD2B COX5A CYP1A2 PML CYP11A1 LOC107984728 ADPGK GOLGA6A NPTN UBL7 MIR6881 NEO1 C15orf59 PPCDC LOC283731 BBS4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |