Assay Details
Target Gene Details
Entrez Gene ID: | 10278 |
Gene Name: | embryonal Fyn-associated substrate |
Gene Aliases: |
CAS3, CASS3, EFS1, EFS2, HEFS, SIN |
Location: |
Chr.14:23356400-23365633 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EFS | NM_001277174.1 | NP_001264103.1 | ||
NM_005864.3 | NP_005855.1 | |||
NM_032459.2 | NP_115835.1 | |||
XM_005267256.1 | XP_005267313.1 | |||
AB001466.1 | BAA24588.1 | |||
AB001467.1 | BAA24589.1 | |||
AK295933.1 | ||||
AK314221.1 | ||||
BC034246.1 | AAH34246.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |