Assay Details
Target Gene Details
Entrez Gene ID: | 100529261 |
Gene Name: | CHURC1-FNTB readthrough |
Gene Aliases: |
- |
Location: |
Chr.14:64914361-65062655 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CHURC1-FNTB | NM_001202558.1 | NP_001189487.1 | ||
NM_001202559.1 | NP_001189488.1 | |||
AK296850.1 | ||||
AK303739.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2748779 | Chr.14:63663324 - 66595809 on Build GRCh38 | Deletion | MIR548H1 ZBTB25 MTHFD1 FNTB GPHN SPTB FUT8-AS1 RAB15 PLEKHG3 SGPP1 MAX LOC102723809 MIR7855 AKAP5 MIR625 PPP1R36 ESR2 SYNE2 TEX21P LINC00238 GPX2 CHURC1 MIR4706 CHURC1-FNTB MIR4708 LOC100506321 LOC100128233 ZBTB1 HSPA2 FUT8 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |