Assay Details
Target Gene Details
Entrez Gene ID: | 282706 |
Gene Name: | DAOA antisense RNA 1 |
Gene Aliases: |
DAOA-AS, DAOAAS, G30 |
Location: |
Chr.13:105459055-105505681 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DAOA-AS1 | NR_040247.1 | |||
AY138548.1 | AAN08434.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv562996 | Chr.13:104426430 - 105664629 on Build GRCh38 | Gain | DAOA DAOA-AS1 |
nsv1048036 | Chr.13:105463559 - 105534128 on Build GRCh38 | Gain | DAOA DAOA-AS1 |