Assay Details
Target Gene Details
Entrez Gene ID: | 2262 |
Gene Name: | glypican 5 |
Gene Aliases: |
- |
Location: |
Chr.13:91398619-92867237 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPC5 | NM_004466.5 | NP_004457.1 | ||
XM_011521054.2 | XP_011519356.1 | |||
XM_011521055.2 | XP_011519357.1 | |||
XM_011521056.2 | XP_011519358.1 | |||
XM_011521057.2 | XP_011519359.1 | |||
XM_011521058.2 | XP_011519360.1 | |||
XM_011521059.2 | XP_011519361.1 | |||
XM_011521060.2 | XP_011519362.1 | |||
XM_017020435.1 | XP_016875924.1 | |||
XM_017020436.1 | XP_016875925.1 | |||
XM_017020437.1 | XP_016875926.1 | |||
AF001462.1 | AAC12261.1 | |||
AK312815.1 | ||||
BC039730.1 | AAH39730.1 | |||
DB100549.1 | ||||
U66033.1 | AAC51118.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |