Assay Details
Target Gene Details
Entrez Gene ID: | 51643 |
Gene Name: | transmembrane BAX inhibitor motif containing 4 |
Gene Aliases: |
CGI-119, GAAP, LFG4, S1R, ZPRO |
Location: |
Chr.12:66136936-66170072 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMBIM4 | NM_001282606.1 | NP_001269535.1 | ||
NM_001282609.1 | NP_001269538.1 | |||
NM_001282610.1 | NP_001269539.1 | |||
NM_016056.3 | NP_057140.2 | |||
AF113127.1 | AAF14868.1 | |||
AF151877.1 | AAD34114.1 | |||
AF161526.1 | AAF29141.1 | |||
AF182041.1 | AAG16898.1 | |||
AF182414.1 | AAG14950.1 | |||
AK075069.1 | BAC11384.1 | |||
AK225158.1 | ||||
AL117550.1 | ||||
BC004401.1 | AAH04401.1 | |||
BG534013.1 | ||||
DC361259.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv526076 | Chr.12:66147526 - 66152320 on Build GRCh38 | Loss | TMBIM4 |
esv3629796 | Chr.12:66130674 - 66161690 on Build GRCh38 | Loss | LLPH TMBIM4 LLPH-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |