Assay Details
Target Gene Details
Entrez Gene ID: | 8630 |
Gene Name: | hydroxysteroid 17-beta dehydrogenase 6 |
Gene Aliases: |
HSE, RODH, SDR9C6 |
Location: |
Chr.12:56752449-56787790 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HSD17B6 | XM_005269207.1 | XP_005269264.1 | ||
XM_005269208.1 | XP_005269265.1 | |||
XM_005269209.1 | XP_005269266.1 | |||
XM_006719672.1 | XP_006719735.1 | |||
XM_011538925.1 | XP_011537227.1 | |||
XM_011538926.1 | XP_011537228.1 | |||
XM_011538927.1 | XP_011537229.1 | |||
DA086202.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss | DCTN2 SPRYD4 MIR1228 CNPY2 ATP5B MBD6 SNORD59A STAT2 LOC101927583 SNORA105C MYO1A NABP2 SDR9C7 MIR6758 OS9 SLC26A10 HSD17B6 ARHGAP9 IL23A PTGES3 TAC3 ZBTB39 RBMS2 NDUFA4L2 SLC39A5 MARS LRP1-AS R3HDM2 GLI1 STAC3 APOF GLS2 NXPH4 LRP1 GPR182 STAT6 ANKRD52 SNORD59B ARHGEF25 TIMELESS B4GALNT1 NEMP1 SHMT2 NACA RDH16 BAZ2A COQ10A PAN2 NAB2 INHBE PIP4K2C INHBC HBCBP DTX3 MIP MIR616 PRIM1 DDIT3 RNF41 KIF5A CS |
nsv826382 | Chr.12:56757680 - 56814823 on Build GRCh38 | Gain | HSD17B6 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |