Assay Details
Target Gene Details
Entrez Gene ID: | 55729 |
Gene Name: | activating transcription factor 7 interacting protein |
Gene Aliases: |
AM, ATF-IP, MCAF, MCAF1, p621 |
Location: |
Chr.12:14365632-14502935 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ATF7IP | NM_001286514.1 | NP_001273443.1 | ||
NM_001286515.1 | NP_001273444.1 | |||
NM_018179.4 | NP_060649.3 | |||
AK025060.1 | ||||
AK299320.1 | ||||
BC063855.1 | AAH63855.1 | |||
DA194386.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv1393n100 | Chr.12:14156211 - 14470137 on Build GRCh38 | Gain |
![]() |
nsv528068 | Chr.12:14117147 - 14382617 on Build GRCh38 | Gain |
![]() |
nsv832335 | Chr.12:14255954 - 14453814 on Build GRCh38 | Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)