Assay Details
Target Gene Details
Entrez Gene ID: | 64375 |
Gene Name: | IKAROS family zinc finger 4 |
Gene Aliases: |
EOS, ZNFN1A4 |
Location: |
Chr.12:56007647-56038435 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Gene Details
Entrez Gene ID: | 105369781 |
Gene Name: | uncharacterized LOC105369781 |
Gene Aliases: |
- |
Location: |
Chr.12:56010092-56025091 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC105369781 | NR_135023.1 | |||
HY183314.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832424 | Chr.12:55881442 - 56073133 on Build GRCh38 | Loss | LOC105369781 IKZF4 SUOX DGKA PMEL CDK2 RPS26 PYM1 RAB5B |
nsv1051961 | Chr.12:55868436 - 56235643 on Build GRCh38 | Gain | LOC105369781 ESYT1 RNF41 CDK2 RPS26 RPL41 NABP2 RAB5B IKZF4 SUOX DGKA SLC39A5 MYL6B PMEL MYL6 ERBB3 LOC107987175 PA2G4 SMARCC2 ZC3H10 PYM1 |
nsv832427 | Chr.12:55998761 - 56118990 on Build GRCh38 | Loss | LOC105369781 IKZF4 SUOX ERBB3 PA2G4 ZC3H10 RPS26 RPL41 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |