Assay Details
Target Gene Details
Entrez Gene ID: | 27345 |
Gene Name: | potassium calcium-activated channel subfamily M regulatory beta subunit 4 |
Gene Aliases: |
- |
Location: |
Chr.12:70366220-70434292 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KCNMB4 | NM_014505.5 | NP_055320.4 | ||
XM_011538188.2 | XP_011536490.1 | |||
AF160967.1 | AAF69805.1 | |||
AF207992.1 | AAF28333.1 | |||
AF215891.1 | AAF75596.1 | |||
BC042446.2 | AAH42446.2 | |||
BC050621.1 | AAH50621.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3580313 | Chr.12:70390534 - 70397851 on Build GRCh38 | Loss | KCNMB4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |