Assay Details
Target Gene Details
Entrez Gene ID: | 8496 |
Gene Name: | PPFIA binding protein 1 |
Gene Aliases: |
L2, SGT2, hSGT2, hSgt2p |
Location: |
Chr.12:27524112-27695564 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv1406n100 | Chr.12:27128236 - 27640822 on Build GRCh38 | Gain | ARNTL2 STK38L SMCO2 PPFIBP1 ARNTL2-AS1 |
dgv2417n54 | Chr.12:27068022 - 27638988 on Build GRCh38 | Gain | ARNTL2 STK38L SMCO2 C12orf71 PPFIBP1 ARNTL2-AS1 |
nsv832358 | Chr.12:27519016 - 27705439 on Build GRCh38 | Gain | REP15 PPFIBP1 |
nsv436825 | Chr.12:27135971 - 27642579 on Build GRCh38 | Insertion | ARNTL2 STK38L SMCO2 PPFIBP1 ARNTL2-AS1 |
esv2745664 | Chr.12:27017516 - 27820117 on Build GRCh38 | Deletion | ARNTL2 KLHL42 STK38L MANSC4 REP15 MRPS35 MED21 SMCO2 C12orf71 PPFIBP1 ARNTL2-AS1 |
esv3628943 | Chr.12:27501670 - 27543029 on Build GRCh38 | Gain | SMCO2 PPFIBP1 |
esv3628940 | Chr.12:27451350 - 27631744 on Build GRCh38 | Gain | SMCO2 PPFIBP1 |
nsv521872 | Chr.12:27114021 - 27635541 on Build GRCh38 | Gain | ARNTL2 STK38L SMCO2 PPFIBP1 ARNTL2-AS1 |
dgv271e199 | Chr.12:27089700 - 27631451 on Build GRCh38 | Deletion | ARNTL2 STK38L SMCO2 PPFIBP1 ARNTL2-AS1 |
More Information
Additional Information:
For this assay, SNP(s) [rs147072311] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |