Assay Details
Target Gene Details
Entrez Gene ID: | 220032 |
Gene Name: | glycerophosphodiester phosphodiesterase domain containing 4 |
Gene Aliases: |
GDE6 |
Location: |
Chr.11:77216558-77301699 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GDPD4 | NM_182833.1 | NP_878253.1 | ||
XM_011544826.1 | XP_011543128.1 | |||
XM_011544833.1 | XP_011543135.1 | |||
XM_011544834.1 | XP_011543136.1 | |||
XM_017017348.1 | XP_016872837.1 | |||
XM_017017349.1 | XP_016872838.1 | |||
XM_017017350.1 | XP_016872839.1 | |||
XM_017017351.1 | XP_016872840.1 | |||
AY313782.1 | AAQ72549.1 | |||
AY326450.1 | AAP92403.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv509434 | Chr.11:77115129 - 77231618 on Build GRCh38 | Insertion | CAPN5 GDPD4 MYO7A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |