Assay Details
Target Gene Details
Entrez Gene ID: | 7003 |
Gene Name: | TEA domain transcription factor 1 |
Gene Aliases: |
AA, NTEF-1, REF1, TCF-13, TCF13, TEAD-1, TEF-1 |
Location: |
Chr.11:12674422-12944737 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TEAD1 | NM_021961.5 | NP_068780.2 | ||
AI199443.1 | ||||
BC115398.1 | ||||
DR000961.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv825769 | Chr.11:12647740 - 12703894 on Build GRCh38 | Gain | TEAD1 |
esv3625395 | Chr.11:12409149 - 13018985 on Build GRCh38 | Gain | TEAD1 RASSF10 PARVA LINC00958 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |