Assay Details
Target Gene Details
Entrez Gene ID: | 9177 |
Gene Name: | 5-hydroxytryptamine receptor 3B |
Gene Aliases: |
5-HT3B |
Location: |
Chr.11:113904796-113949682 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HTR3B | NM_006028.4 | NP_006019.1 | ||
XM_011543063.1 | XP_011541365.1 | |||
XM_017018552.1 | XP_016874041.1 | |||
AF080582.1 | AAD12242.1 | |||
AF169255.1 | AAF03691.1 | |||
AK314268.1 | ||||
AM293589.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832273 | Chr.11:113784031 - 113957584 on Build GRCh38 | Loss | USP28 LOC390251 HTR3B |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |