Assay Details
Target Gene Details
Entrez Gene ID: | 1500 |
Gene Name: | catenin delta 1 |
Gene Aliases: |
CAS, CTNND, P120CAS, P120CTN, p120, p120(CAS), p120(CTN) |
Location: |
Chr.11:57761762-57819180 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Gene Details
Entrez Gene ID: | 100528016 |
Gene Name: | TMX2-CTNND1 readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.11:57712523-57819180 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMX2-CTNND1 | NR_037646.1 | |||
AB002382.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv327 | Chr.11:57765169 - 57783274 on Build GRCh38 | Deletion | CTNND1 TMX2-CTNND1 |
nsv508633 | Chr.11:57652112 - 57792573 on Build GRCh38 | Deletion | C11orf31 TMX2 BTBD18 CLP1 CTNND1 ZDHHC5 TMX2-CTNND1 MED19 |
nsv832164 | Chr.11:57640144 - 57811353 on Build GRCh38 | Loss | C11orf31 TMX2 BTBD18 MIR130A CLP1 CTNND1 ZDHHC5 TMX2-CTNND1 YPEL4 MED19 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |