Assay Details
Target Gene Details
Entrez Gene ID: | 84188 |
Gene Name: | fatty acyl-CoA reductase 1 |
Gene Aliases: |
MLSTD2, PFCRD, SDR10E1 |
Location: |
Chr.11:13668659-13732346 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FAR1 | NM_032228.5 | NP_115604.1 | ||
XM_011520400.1 | XP_011518702.1 | |||
AK026381.1 | BAB15467.1 | |||
AK315527.1 | ||||
AY358784.1 | AAQ89144.1 | |||
AY423606.1 | AAR84086.1 | |||
AY600449.1 | AAT42129.1 | |||
BC017377.2 | AAH17377.1 | |||
CR936619.1 | CAI56762.1 | |||
DA515509.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |